missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ MEP1A Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse MEP1A (NP_032611.2) (Met 1-Arg 615) fused with a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50057M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15312616

  • £132.21 / 5µg


Estimated Shipment
14-06-2018
Add to basket

Includes: One vial of lyophilized protein

Description

Description

Meprin 1 alpha (MEP1A), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave a fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH; the specific activity is >400pmole/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
69kDa
5μg
Proteases
AI098089, AW107200, Mep-1, Mep-1a, Mep1
MEP1A
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Meprin A, alpha (PABA peptide hydrolase)
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.