missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ MAP1D Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human METAP1D (NP_954697.1) (Arg 44-Ala 335) with a polyhistidine tag at the C-terminus and an initial Met an the N-terminus

Brand:  Thermo Scientific™ 10883H08B5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15297325

  • £93.43 / 5µg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

Methionyl aminopeptidase type 1D (mitochondrial) (METAP1D), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measure by its ability to remove methionine from a fluorogenic peptide substrate H-Met-Gly-Pro-AMC; the resulting Gly-Pro-AMC is cleaved by human DPP4/CD26, the specific activity is >30pmole/min./μg
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
33.4kDa
5μg
Proteases
MAP1D
MAP1D
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Methionyl aminopeptidase type 1D (mitochondrial)
Assay Standard
Insect
Videos

For Research Use Only. Not for use in diagnostic procedures.