missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ MAG Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human MAG (P20916) (Met 1-Pro 516) was fused with a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 13186H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15216225

  • £216.53 / 50µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Myelin Associated Glycoprotein (MAG), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
56kDa
50μg
Other Proteins|Neuropeptides
GMA, S-MAG, SIGLEC-4A, SIGLEC4A
MAG
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Myelin associated glycoprotein
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.