missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ LTA Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of human LTA (P01374) (Leu 35-Leu 205) fused to an initial Met at the N-terminus

Brand:  Thermo Scientific™ 10270HNAE25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15235055

  • £191.85 / 25µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Lymphotoxin Alpha (TNF Superfamily Member 1) (LTA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cytotoxicity assay using L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D, the ED50 for this effect is typically 0.1-0.4ng/mL
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
18.8kDa
5 x 5μg
TNF (Tumor Necrosis Factor)
Lymphotoxin alpha (TNF superfamily, member 1)
Assay Standard
E. coli
Lyophilized
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
LTA, DAMA-25N12, 13-004, LT, TNFB, TNFSF1
LTA
Cytokines & Receptors
Untagged
Videos

For Research Use Only. Not for use in diagnostic procedures.