missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ LCN2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human Lipocalin 2 (LCN2) precursor (NP_005555.2) (Met 1-Gly 198) fused to the polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10222H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15264935

  • £125.97 / 25µg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

Lipocalin 2 (LCN2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind Iron(III) dihydroxybenzoic acid (Fe(DHBA)3); the binding of Fe(DHBA)3 results in the quenching of Trp fluorescence in recombinant human Lipocalin-2
    Recombinant human Lipocalin-2 can bind >1.5μM of Fe(DHBA)3
  • Formulation: Lyophilized in 25mM MES, 150mM NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
22kDa
5 x 5μg
Other Proteins
NGAL, LCN2
LCN2
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Lipocalin 2
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.