missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ LAIR2 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human LAIR2 (NP_002279.2) (Met 1-Pro 152)

Brand:  Thermo Scientific™ 10868HNAH25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15217315

  • £127.01 / 25µg

Estimated Shipment
Add to basket



Leukocyte-Associated Immunoglobulin-Like Receptor 2 (LAIR2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of HT-29 human colon adenocarcinoma cells; when 5 x 104 cells/well are added to recombiannt human LAIR2 coated plates (50μg/mL with 100μL/well), approximately 30% will adhere after 30 minutes at 37°C (optimal concentration depends on cell type as well as the application or research objectives)
  • Formulation: Lyophilized in 50mM Tris, 0.7M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5 x 5μg
Other Proteins
LAIR2, XXbac-BCX535A19.6, CD306, mgC71634
CD Proteins
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Leukocyte-associated immunoglobulin-like receptor 2
Assay Standard

For Research Use Only. Not for use in diagnostic procedures.