missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ KNG1 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse KNG1 isoform 3 (NP_001095882.1) (Glu 21-Ser 480) fused to a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus

Brand:  Thermo Scientific™ 50474M08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15333286

  • £2748.24 / 250µg


Estimated Shipment
14-06-2018
Add to basket

Includes: Five vials of lyophilized protein

Description

Description

Kininogen 1 (KNG1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to inhibit papain cleavage of a fluorogenic peptide substrate Z-FR-AMC, the IC50 value is <5nM
  • Formulation: Lyophilized in 20mM Tris, 100mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
52.5kDa
5 x 50μg
Other Proteins
Kng, mgC123437, Kng1
KNG1
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Kininogen 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.