missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ KLK7 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human KLK7 (NP_005037.1) extracellular domain (Met 1-Arg 253) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10416H08H5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15275375

  • £142.62 / 5µg


Estimated Shipment
05-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Kallikrein-Related Peptidase 7 (KLK7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK (Dnp) NH2, (R&D Systems Cat. No. ES002), the specific activity is >150pmoles/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
26.7kDa
5μg
Proteases
PRSS6, SCCE, KLK7
KLK7
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Kallikrein-related peptidase 7
Assay Standard
Mammalian
Videos