missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ KLK4 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human KLK4 (NP_004908.3) (Met 1-Ser 254) fused to the polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11857H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15225575

  • £142.62 / 5µg


Estimated Shipment
13-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Kallikrein-related peptidase 4 (KLK4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate Boc-VPR-AMC; the specific activity is >250pmole/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
25.8kDa
5μg
Proteases
AI2A1, ARM1, EMSP, EMSP1, KLK-L1, mgC116827, mgC116828, PRSS17, PSTS
KLK4
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Kallikrein-related peptidase 4
Assay Standard
Mammalian
Videos