missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ KITL Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse KITL (P20826-1) (Met 1-Ala 189) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50487M08B5

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 13433149

  • £86.93 / 250µg


Estimated Shipment
14-06-2018
Add to basket

Includes: One vial of lyophilized protein

Description

Description

KIT ligand (KITL), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: 1. Measured by its binding ability in a functional ELISA; immobilized mouse KITL at 2μg/mL (100μL/well) can bind mouse KIT with a linear ranger of 1.28-32ng/mL; 2. Measured in a cell proliferation assay using TF-1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323), the EC50 for this effect is typically 0.5-5ng/mL
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
19.7kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Kit ligand
Assay Standard
Insect
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Growth Factors
Clo, Con, Gb, Kitlg, mgf, SCF, SF, SLF, Sl, Steel, c ontrasted
KITL
Growth Factors & Receptors
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.