missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ KIT (c-Kit) Recombinant Human Protein (without Catalytic Activity Domain), His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human KIT isoform 2 (P10721-2) (Met 1-Thr 516) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50530M02H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13456316

  • £56.74 / 5µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Kit oncogene (KIT/c-Kit), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human KIT at 20μg/mL (100μL/well) can bind biotinylated mouse KITL with a linear range of 1.28-6.4ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
16590
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Receptor Tyrosine Kinases|Cytokine Receptors
Bs, CD117, Fdc, Gsfsco1, Gsfsco5, Gsfsow3, SCO1, SCO5, SOW3, Ssm, Tr-kit, W, c-KIT
KIT
Growth Factors & Receptors/CD Proteins
PDGFR Kinase Family
Lyophilized
82.5kDa
5μg
Truncated/Recombinant
<1 EU/μg
KIT oncogene
Assay Standard
Mammalian
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.