missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ KDR (VEGFR2) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human KDR (NP_002244.1) (Met 1-Glu 764) fused with a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10012H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13492909

  • £55.70 / 5µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Kinase insert domain receptor (KDR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Using the Octet RED System; the affinity constant (Kd) of human KDR-His bound to biotinylated human NRP2 (VEGF165) was 5.9nM
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
84.6kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Kinase insert domain receptor (a type III receptor tyrosine kinase)
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
VEGF (Vascular Endothelial Growth Factor)
CD309, FLK1, VEGFR, VEGFR2, KDR
KDR
Kinases & Inhibitors/Growth Factors & Receptors/CD Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.