Sino Biological™ JAM3 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse JAM3 (NP_075766.1) extracellular domain (Met 1-Asn 241) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 50465M08H5

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Includes: One vial of protein in suspension buffer

Product Code. 15323276

Quantity Price
1 £ 63.0 / 5µg
Estimated Shipment date
from Supplier 18-04-2017
Add to basket


Description and Specification

Specification

Purity ≥90% by SDS-PAGE
Protein Subtype Intercellular Adhesion Molecule
Validated Application Assay Standard
Termination C-Terminal
Molecular Weight 25kDa
Format Lyophilized
Endotoxin Concentration <1 EU/μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Common Name Junction adhesion molecule 3
Purification Method PAGE
Protein Tag His Tag
Gene Symbol JAM3
Structural Form Truncated/Recombinant
Gene ID (Entrez) 83964
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Gene Alias 1110002N23Rik, C85515, JAM-3, JAM-C, Jcam3, Jam3
Protein Family Other Proteins
Quantity 5μg
Expression System Mammalian

Junction Adhesion Molecule 3 (JAM3), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris-Citrate, 300mM NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.