missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ JAM2 Recombinant Mouse Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse JAM2 (NP_076333.3) extracellular domain (Met 1-Asn 236) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50464M02H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15323266

  • £127.01 / 25µg


Estimated Shipment
14-06-2018
Add to basket

Includes: Five vials of lyophilized protein

Description

Description

Junction Adhesion Molecule 2 (JAM2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of Jurkat human leukemic T cells; when 8 x 104 cells/well are added to JAM2 coated plates (0.2μg/mL and 100μL/well), approximately 40-55% will adhere specifically after 60 minutes at 37°C
  • Formulation: Lyophilized in 40mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
50.3kDa
5 x 5μg
Intercellular Adhesion Molecule
RP23-253E17.2, 1110002N23Rik, 2410030G21Rik, 2410167M24Rik, AU016127, JAM-2, JAM-B
JAM2
CD Proteins
IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Junction adhesion molecule 2
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.