missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ JAG1 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the human JAG1 (NP_000205.1) extracellular domain (Met 1-Ser 1046) fused to Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 11648H02H25

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15228775

  • £125.97 / 25µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Jagged 1 (JAG1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
182
PAGE
5 x 5μg
C-Terminal
Other Proteins
JAG1, AGS, AHD, AWS, CD339, HJ1, JAGL1, mgC104644
JAG1
CD Proteins
IgG1 Fc Tag
Lyophilized
137kDa
>85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
<1 EU/μg
Jagged 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.