missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ISG15 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of human ISG15 (AAH09507.1) (Met 1-Gly157)

Brand:  Thermo Scientific™ 12729HNAE5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15256045

  • £47.65 / 5µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

ISG15 Ubiquitin-Like Modifier (ISG15), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
9636
PAGE
5μg
Full Length/Recombinant
<1 EU/μg
ISG15 ubiquitin-like modifier
Assay Standard
E. coli
Lyophilized
17.9kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Other Cytokines
G1P2, IFI15, IP17, UCRP, hUCRP
ISG15
Other Proteins
Videos

For Research Use Only. Not for use in diagnostic procedures.