Sino Biological™ ISG15 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of human ISG15 (AAH09507.1) (Met 1-Gly157)

Overview
Brand: Invitrogen

Manufacturer Part Number: 12729HNAE250

RECOM HU ISG15

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15246045

Quantity Price
1 £ 87.25 / 250µg
EU Stock 3
Add to basket


Description and Specification

Specification

Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5 x 50μg
Gene Symbol ISG15
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 17.9kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias G1P2, IFI15, IP17, UCRP, hUCRP
Common Name ISG15 ubiquitin-like modifier
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
Format Lyophilized
Gene ID (Entrez) 9636

ISG15 Ubiquitin-Like Modifier (ISG15), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.