missing translation for 'onlineSavingsMsg'
Learn More

INSR, Short Isoform, Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 944) of human INSR short isoform (NP_001073285.1) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 11086H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13446096

  • £144.41 / 25µg

Estimated Shipment date
from Supplier 03-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Insulin receptor (INSR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; when 1μg/mL of biotinylated human insulin is immobilized onto a streptavidin coated plate, it can bind human insulin receptor (INSR) with a linear range of 0.3-40μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3643
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Hormone Receptors
CD220, HHF5, INSR
INSR
Hormones & Receptors
Insulin & IGF Receptor Kinase Family
Lyophilized
106.0kDa
5 x 5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Insulin receptor, short isoform
Assay Standard
Mammalian
His Tag
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.