missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ INSR Recombinant Human Protein (Long Isoform, without Catalytic Activity Domain), His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 956) of human INSR long isoform (NP_000199.2) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11081H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13496086

  • £238.39 / 50µg


Estimated Shipment
14-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Insulin receptor (INSR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; when 1μg/mL of biotinylated human insulin is immobilized onto a streptavidin coated plate, it can bind human INSR, isoform long, with a linear range of 0.625-10μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3643
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Hormone Receptors
CD220, HHF5, INSR
INSR
Growth Factors & Receptors
Insulin & IGF Receptor Kinase Family
Lyophilized
107.0kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Insulin receptor, long isoform
Assay Standard
Mammalian
His Tag
Videos