missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL6ST Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse extracellular domain (Met 1-Glu 617) of IL6ST (NP_034690.3) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50135M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13496236

  • £56.74 / 5µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Interleukin 6 signal transducer (IL6ST), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: 1. Measured by its ability to bind mouse IL11RA in a functional ELISA; 2. Measured by its ability to inhibit the IL6R enhancement of IL6 activity on M1 mouse myeloid leukemia cells (Saito, T. et al. 1991, J. Immunol. 147:168), the ED50 for this effect is typically 0.2-0.8μg/mL in the presence of 50ng/mL recombinant human IL6sR and 100ng/mL recombinant human IL6
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
16195
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
IL6ST, 5133400A03Rik, AA389424, BB405851, CD130, D13Ertd699e, gp130
IL6ST
Cytokines & Receptors/CD Proteins
His Tag
Lyophilized
68kDa
5μg
Recombinant
<1 EU/μg
Interleukin 6 signal transducer
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.