missing translation for 'onlineSavingsMsg'
Learn More

IL6ST Recombinant Mouse Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 617) of mouse IL6ST (NP_034690.3) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Invitrogen 50135M03H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13436236

  • £147.59 / 25µg

Estimated Shipment
12-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Interleukin 6 signal transducer (IL6ST), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: Measured by its ability to inhibit the IL6RA enhancement of IL6 activity on M1 mouse myeloid leukemia cells (Saito, T. et al. 1991, J. Immunol. 147:168); the ED50 for this effect is typically 0.05-0.2μg/mL in the presence of 50ng/mL recombinant human IL6sR and 100ng/mL recombinant human IL6
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
16195
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
IL6ST,5133400A03Rik, AA389424, BB405851, CD130, D13Ertd699e, gp130
IL6ST
Cytokines & Receptors/CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
94.7kDa
5 x 5μg
Truncated/Recombinant
<1 EU/μg
Interleukin 6 signal transducer
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.