missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL6RA Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse IL6RI± (NP_034689.2) extracellular domain (Ile 44-Glu 357) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 50280M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15383056

  • £84.07 / 5µg


EU Stock 2
Add to basket

Includes: One vial of protein in suspension buffer

Description

Description

Interleukin 6 Receptor Alpha (IL6RA), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
16194
PAGE
5μg
C-Terminal
<1 EU/μg
Interleukin 6 receptor alpha
Assay Standard
Mammalian
Lyophilized
39kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
Il6ra, CD126, IL-6R, Il6r, mgC30256
IL6RA
Chemokines & Receptors/CD Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.