missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL6R Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 365) of human IL6R (NP_000556.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10398H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465866

  • £201.96 / 25µg


Estimated Shipment
26-07-2018
Add to basket

Description

Description

Interleukin 6 receptor (IL6R), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human IL6 at 8μg/mL (100μL/well) can bind recombinant human IL6R with a linear range of 1.25-20.0ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3570
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
IL6R, CD126, IL-6R-1, IL-6R-alpha, IL6RA, mgC104991
IL6R
Cytokines & Receptors/CD Proteins
His Tag
Lyophilized
40kDa
5 x 5μg
Truncated/Recombinant
<1 EU/μg
Interleukin 6 receptor
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.