missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL4 Recombinant Human Protein, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL4 isoform 1 (P05112-1) (His 25-Ser 153) retaining the initial Met
£66.75 - £345.00
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Molecular Weight (g/mol) | 15kDa |
Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Structural Form | Full Length/Recombinant |
Gene Alias | BCGF-1, BCGF1, BSF1, IL-4, mgC79402 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13443069
|
Invitrogen
11846HNAE50 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13433069
|
Invitrogen
11846HNAE5 |
5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Interleukin 4 (IL-4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.- Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al.,1989, J. Cell Physiol. 140:323.); the EC50 for this effect is typically 0.1-0.3ng/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
BCGF-1, BCGF1, BSF1, IL-4, mgC79402 | |
IL4 | |
E. coli | |
Lyophilized | |
≥90% by SDS-PAGE |
15kDa | |
Full Length/Recombinant | |
Interleukin 4 | |
Untagged | |
Cytokines & Receptors | |
Interleukins |
For Research Use Only. Not for use in diagnostic procedures.