missing translation for 'onlineSavingsMsg'
Learn More

IL4 Recombinant Human Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL4 isoform 1 (P05112-1) (His 25-Ser 153) retaining the initial Met

Brand:  Invitrogen 11846HNAE50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13443069

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Interleukin 4 (IL-4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al.,1989, J. Cell Physiol. 140:323.); the EC50 for this effect is typically 0.1-0.3ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
50μg
Full Length/Recombinant
Interleukin 4
Untagged
Cytokines & Receptors
Interleukins
15kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
BCGF-1, BCGF1, BSF1, IL-4, mgC79402
IL4
E. coli
Lyophilized
≥90% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.