Learn More
IL4 Recombinant Human Protein, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL4 isoform 1 (P05112-1) (His 25-Ser 153) retaining the initial Met
Brand: Invitrogen 11846HNAE5
Code : NEW
Additional Details : Weight : 0.00001kg
Description
Interleukin 4 (IL-4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.- Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al.,1989, J. Cell Physiol. 140:323.); the EC50 for this effect is typically 0.1-0.3ng/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
5μg | |
Full Length/Recombinant | |
Interleukin 4 | |
Untagged | |
Cytokines & Receptors | |
Interleukins |
15kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
BCGF-1, BCGF1, BSF1, IL-4, mgC79402 | |
IL4 | |
E. coli | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.