missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL37 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL1F7 isoform A (Q9NZH6-2) (Lys 27-Asp 192) fused to an initial Met

Brand:  Thermo Scientific™ 10155HNAE250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15204765

  • £1251.29 / 250µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Interleukin 37 (IL37), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
27178
PAGE
5 x 50μg
Full Length/Recombinant
IL37, FIL1, FIL1(ZETA), FIL1Z, IL-1F7, IL-1H, IL-1H4, IL-1RP1, IL-37, IL1F7, IL1H4, IL1RP1
IL37
Cytokines & Receptors
Lyophilized
18.7kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Interleukin 37
Assay Standard
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.