missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL36B (IL1F8) Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL36B isofrom 2 (Q9NZH7-2) (Met 1-Glu 157)

Brand:  Thermo Scientific™ 10579HNAE50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15286955

  • £354.99 / 50µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

Interleukin 36 Beta (IL36B), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
27177
PAGE
50μg
Full Length/Recombinant
IL36B, FIL1, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1F8, IL-1H2, IL1-ETA, mgC126880, mgC126882
IL36B
Cytokines & Receptors
Lyophilized
17.7kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Interleukin 36 beta
Assay Standard
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.