missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL36A (IL1F6) Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL1F6 (Q9UHA7) (Met 1-Phe 158)

Brand:  Thermo Scientific™ 10607HNAE50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15246985

  • £354.99 / 50µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Interleukin 36 alpha (IL36A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
27179
PAGE
50μg
Full Length/Recombinant
IL36A, FIL1, FIL1(EPSILON), FIL1E, IL-1F6, IL1(EPSILON), IL1F6, mgC129552, mgC129553
IL36A
Cytokines & Receptors
Lyophilized
17.7kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Interleukin 36 alpha
Assay Standard
E. coli
Videos