missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL33 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the C-terminal segment of human IL33 (NP_002296.1) (Ser 112-Thr 270)

Brand:  Thermo Scientific™ 10368HNAE25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15295265

  • £303.98 / 25µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Interleukin 33 (IL33), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL33 at 10μg/mL (100μg/well) can bind human IL1RL1 with a linear range of 0.31-5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
20kDa
5 x 5μg
Interleukins
Interleukin 33
Assay Standard
E. coli
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
C9orf26, DKFZp586H0523, DVS27, NF-HEV, NFEHEV, RP11-575C20.2
IL33
Cytokines & Receptors
Untagged
Videos