missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL2RG Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human IL2RG (P31785) extracellular domain (Met 1-Asn 254) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10555H08B5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15216915

  • £74.43 / 5µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

Interleukin 2 Receptor Gamma (IL2RG), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind biotinylated recombinant rat IL2 in a functional ELISA
  • Formulation: 20mM Tris, 500mM NaCl, pH 7.4, 10% gly
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
28.8kDa
5μg
Interleukins|Cytokine Receptors
CD132, CIDX, IL-2RG, IMD4, P64, SCIDX, SCIDX1
IL2RG
Insect
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Interleukin 2 receptor gamma
Assay Standard
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.