missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ IL2RB Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 239) of human IL2RB (NP_000869.1) fused to the C-terminal fused Fc region of human IgG1

Brand:  Invitrogen 10696H02H25

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15297075

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Interleukin 2 Receptor beta (IL2RB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind biotinylated recombinant rat IL2 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
51.6kDa
5 x 5μg
Interleukins|Cytokine Receptors
IL2RB, RP5-1170K4.6, CD122, P70-75
IL2RB
Cytokines & Receptors
IgG1 Fc Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Interleukin 2 receptor beta
Assay Standard
Mammalian