missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL2RA Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human IL2 receptor ± (NP_000408.1) (Met 1-Cys 213) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10165H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15274785

  • £158.24 / 25µg


Estimated Shipment
12-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Interleukin 2 Receptor Alpha (IL2RA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by Octet RED System, the affinity constant (Kd) of Human IL2RA-his (Mfr. No. 10165-H08H) bound to Human IL2 was 35nM
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3559
PAGE
5 x 5μg
C-Terminal
<1 EU/μg
Interleukin 2 receptor alpha
Assay Standard
Mammalian
Lyophilized
23.3kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
RP11-536K7.1, CD25, IDDM10, IL2R, TCGFR
IL2RA
Cytokines & Receptors/CD Proteins
His Tag
Videos