missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL2RA (CD25) Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the amino acid residues (Met 1-Cys 213) of the human IL2RA precursor (NP_000408.1) fused to the human IgG1 Fc region at the C-terminus

Brand:  Thermo Scientific™ 10165H02H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13485746

  • £55.70 / 5µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

Interleukin 2 receptor, alpha (IL2RA/CD25), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3559
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
RP11-536K7.1, CD25, IDDM10, IL2R, TCGFR
IL2RA
Cytokines & Receptors/CD Proteins
IgG1 Fc Tag
Lyophilized
48.5kDa
5μg
Recombinant
<1 EU/μg
Interleukin 2 receptor, alpha
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.