missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL27RA Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 514) of human IL27RA (NP_004834.1) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10489H02H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13425886

  • £144.70 / 25µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Interleukin 27 receptor, alpha (IL27RA), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL27 at 20μg/mL (100μL/well) can bind human IL27RA with a linear range of 0.064-1.6μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
9466
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
IL27RA, CRL1, IL27R, TCCR, WSX1, zcytor1
IL27RA
Cytokines & Receptors
IgG1 Fc Tag
Lyophilized
79kDa
5 x 5μg
Truncated/Recombinant
<1 EU/μg
Interleukin 27 receptor, alpha
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.