missing translation for 'onlineSavingsMsg'
Learn More

IL21R Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human IL21R (Q9HBE5-1) extracellular domain (Met 1-Pro 236), with a polyhistidine tag at the C-terminus

Brand:  Invitrogen 11483H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13446136

  • £147.00 / 25µg

Estimated Shipment
20-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Interleukin 21 receptor (IL21R), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind human IL21 in a functional ELISA; 2. Measured by its ability to inhibit IL21-induced Interferon gamma secretion by human natural killer lymphoma NK-92 cells, the ED50 for this effect is typically 1-5ng/mL in the presence of 100ng/mLof recombinant human IL21; 3. Measured by its ability to inhibit IL21 induced Interferon gamma secretion by human natural killer lymphoma NK-92 cells, the ED50 for this effect is typically 50-200pg/mL in the presence of 10ng/mL of recombinant human IL21
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Interleukins|Cytokine Receptors
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
50615
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Interleukin 21 receptor
His Tag
Cytokines & Receptors
Assay Standard
Lyophilized
26.5kDa
5 x 5μg
Truncated/Recombinant
UNQ3121/PRO10273, mgC10967, NILR
IL21R
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.