Learn More
IL2 Recombinant Rat Protein, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the rat IL2 (P17108) (Ala 21-Gln 155) fuse to an initial Met at the N-terminus
Brand: Invitrogen 80075RNAE50
Code : NEW
Additional Details : Weight : 0.01000kg
Includes: One vial of lyophilized protein
Description
Interleukin 2 (IL2), recombinant rat protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.- Activity: This recombinant protein has not been tested
- Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
15.6kDa | |
50μg | |
Full Length/Recombinant | |
Interleukin 2 | |
Untagged | |
Cytokines & Receptors | |
≥90% by SDS-PAGE |
116562 | |
PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
IL-2, T-cell growth factor, TCGF, interleukin-2 | |
IL2 | |
E. coli | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.