missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL2 Recombinant Rat Protein

This recombinant protein was expressed from a DNA sequence encoding the rat IL2 (P17108) (Ala 21-Gln 155) fuse to an initial Met at the N-terminus

Brand:  Thermo Scientific™ 80075RNAE5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15309586

  • £59.34 / 5µg


Estimated Shipment
06-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: One vial of lyophilized protein

Description

Description

Interleukin 2 (IL2), recombinant rat protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
116562
PAGE
5μg
Full Length/Recombinant
Interleukin 2
Assay Standard
E. coli
Lyophilized
15.6kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
IL-2, T-cell growth factor, TCGF, interleukin-2
IL2
Cytokines & Receptors
Untagged
Videos