Sino Biological™ IL1RN Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL1RA isoform1 (NP_776214.1) (Arg 26-Glu 177)

Overview
Brand: Invitrogen

Manufacturer Part Number: 10123HNAE25

RECOM HU IL1RN

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15234695

Quantity Price
1 £ 73.5 / 25µg
EU Stock 1
Add to basket


Description and Specification

Specification

Molecular Weight 17.3kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Structural Form Full Length/Recombinant
Protein Subtype Interleukins
Endotoxin Concentration <1 EU/μg
Gene Alias IL1RN, ICIL-1RA, IL-1ra3, IL1F3, IL1RA, IRAP, mgC10430
Common Name Interleukin 1 receptor antagonist
Gene Symbol IL1RN
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 3557

Interleukin 1 Receptor Antagonist (IL1RN), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.