missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL1RL1 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the human IL1RL1, isoform 2 (NP_003847.2), residues 1 to 328, fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10105H02H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13405706

  • £55.70 / 5µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

Interleukin 1 receptor-like 1 (IL1RL1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL33 at 10μg/mL (100μL/well) can bind human IL1RL1-Fc with a linear range of 0.31-5ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
9173
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
IL-1RL1, T1, ST2, DER4, ST2L, ST2V, FIT-1, mgC32623
IL1RL1
Cytokines & Receptors
IgG1 Fc Tag
Lyophilized
67.1kDa
5μg
Recombinant
<1 EU/μg
Interleukin 1 receptor-like 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.