missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL1RL1 Recombinant Human Protein, DDDDK Tag

This recombinant protein was expressed from a DNA sequence encoding the secreted isoform 1 (Met 1-Phe 328) of human IL1RL1 (NP_003847.2) fused to a DDDDK tag at the C-terminus

Brand:  Thermo Scientific™ 10105HCCH50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13455706

  • £186.34 / 50µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Interleukin 1 receptor-like 1 (IL1RL1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
9173
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
IL-1RL1, T1, ST2, DER4, ST2L, ST2V, FIT-1, mgC32623
IL1RL1
Cytokines & Receptors
DDDDK
Lyophilized
35.6kDa
50μg
Recombinant
<1 EU/μg
Interleukin 1 receptor-like 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.