missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL1RAPL2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 356) of human IL1RAPL2 (NP_059112.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10156H08H250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15244765

  • £464.29 / 250µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Interleukin 1 receptor accessory protein-like 2 (IL1RAPL2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability to inhibit IL1B dependent proliferation in D10.G4.1 mouse helper T cells; approximately 5-20μg/mL will inhibit 50% of the biological response due to 40pg/mL of IL1B
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
40.7kDa
5 x 50μg
NFκB Pathway Proteins|Cytokine Receptors
IL1R9, IL-1R9, TIGIRR-1, IL1RAPL-2
IL1RAPL2
Cytokines & Receptors
His Tag
Lyophilized
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Interleukin 1 receptor accessory protein-like 4
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.