missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL1R2 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 343) of human IL1R2 (NP_004624.1) fused to the human IgG1 Fc region at the C-terminus

Brand:  Thermo Scientific™ 10111H02H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465716

  • £56.74 / 5µg


Estimated Shipment
05-09-2018
Add to basket

Description

Description

Interleukin 1 receptor, type II (IL1R2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human IL1B at 2μg/mL (100μL/well) can bind recombinant human IL1R2-Fc with a linear range of 6.25-400ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
7850
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
CD121b, IL1RB, mgC47725, IL1R2
IL1R2
Cytokines & Receptors/CD Proteins
IgG1 Fc Tag
Lyophilized
64.5kDa
5μg
Truncated/Recombinant
<1 EU/μg
Interleukin 1 receptor, type II
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.