missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL1F5 Recombinant Mouse Protein

This recombinant protein was expressed from a DNA sequence encoding the mouse IL1F5 (NP_062324.2) (Met 2-Asp 156)

Brand:  Thermo Scientific™ 50213MNAE5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15322946

  • £83.55 / 5µg


Estimated Shipment
11-06-2018
Add to basket

Includes: One vial of protein in suspension buffer

Description

Description

Interleukin 1 Family Member 5 (Delta) (IL1F5), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
54450
PAGE
5μg
Full Length/Recombinant
<1 EU/μg
Interleukin 1 family member 5 (delta)
Assay Standard
E. coli
Lyophilized
17kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Interleukins
RP23-176J12.6, AI413231, Fil1delta, Il-1h3, Il1hy1
IL1F5
Cytokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.