missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ IL1B Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the proform of human IL1B (NP_000567.1) (Met 1-Ser 269) fused to a polyhistidine tag at the N-terminus

Brand:  Invitrogen 10139H07E50

UNSPSC : 12352202

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15204725

  • £364.00 / 50µg


Estimated Shipment
09-08-2017
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Interleukin 1, beta (IL1B), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL1B at 10μg/mL (100μL/well) can bind human IL1R1, the EC50 of human IL1R1 is 0.198μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
32.3kDa
50μg
Interleukins
Interleukin 1, beta
Assay Standard
E. coli
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
IL-1, IL1-BETA, IL1F2, IL1B
IL1B
Cytokines & Receptors
His Tag