missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL1B Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL1B (NP_000567.1) (Ala 117-Ser 269)

Brand:  Thermo Scientific™ 10139HNAE250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15214725

  • £1145.10 / 250µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Interleukin 1, beta (IL1B), a recombinant human protein, is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as immunogens, protein standards, and in other cell biology research applications.
  • Activity: 1.Measured by its binding ability in a functional ELISA; a. Immobilized recombinant human IL1B at 2μg/mL (100μL/well) can bind recombinant human IL1R2 with a linear range of 6.25-400ng/mL; b. Immobilized human IL1B at 10μg/mL (100μL/well) can bind human IL1R1, the EC50 of human IL1R1 is 0.48μg/mL; 2. Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells, the ED50 for this effect is typically 0.5-5pg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3553
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukin 1, beta
Assay Standard
E. coli
Lyophilized
17kDa
5 x 50μg
IL-1, IL1-BETA, IL1F2, IL1B
IL1B
Cytokines & Receptors
Untagged
Videos

For Research Use Only. Not for use in diagnostic procedures.