missing translation for 'onlineSavingsMsg'
Learn More

IL1B Recombinant Cynomolgus Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mature form of Cynomolgus (Macaca fascicularis) IL1B (Ala 117-Ser 269) fused to an initial Met

Brand:  Invitrogen 90010CNAE5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15349656

  • £112.43 / 5µg


Estimated Shipment
14-01-2019
Add to basket

Includes: One vial of protein in suspension buffer

Description

Description

Interleukin 1 Beta (IL1B), recombinant cynomolgus protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells; the ED50 for this effect is typically 5-40pg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
704701
PAGE
5μg
Full Length/Recombinant
<1 EU/μg
Interleukin 1 beta
Assay Standard
E. coli
Lyophilized
17.5kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Interleukins
IL1B
IL1B
Cytokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.