missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL1A Recombinant Mouse Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse IL1A (NP_034684.2) (Ser 115-Ser 270), with an initial Met at the N-terminus

Brand:  Thermo Scientific™ 50114MNAE250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15352746

  • £1471.98 / 250µg


Estimated Shipment
02-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

Description

Description

Interleukin 1 alpha (IL1A), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: 1. Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells, the ED50 for this effect is typically 2-10pg/mL; 2. Measured by its binding ability in a functional ELISA; immobilized mouse IL1A at 10μg/mL (100μL/well) can bind mouse IL1R1, the EC50 of mouse IL1R1 is 0.13μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
17kDa
5 x 50μg
Interleukins
Interleukin 1 alpha
Assay Standard
E. coli
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
RP23-160G19.8, Il-1a
IL1A
Cytokines & Receptors
Untagged
Videos