missing translation for 'onlineSavingsMsg'
Learn More

IL18RAP Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding a fragment of human IL18RAP (NP_003844.1), residues 1 to 357, fused to the Fc region of human IgG1 at the C-terminus

£57.90 - £450.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL18RAP, IL-1R7, ACPL, CD218b, CDw218b, IL18RB, mgC120589, mgC120590
Gene ID (Entrez) 8807
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13465756 View Documents Invitrogen
10176H02H5
5μg £57.90
5µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 18-06-2020
Please to purchase this item. Need a web account? Register with us today!
13445756 View Documents Invitrogen
10176H02H25
5 x 5μg £148.00
25µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 18-06-2020
Please to purchase this item. Need a web account? Register with us today!
13475756 View Documents Invitrogen
10176H02H50
50μg £191.00
50µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 18-06-2020
Please to purchase this item. Need a web account? Register with us today!
13455756 View Documents Invitrogen
10176H02H250
5 x 50μg £450.00
250µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 18-06-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Interleukin 18 receptor accessory protein (IL18RAP), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
8807
<1 EU/μg
IL18RAP, IL-1R7, ACPL, CD218b, CDw218b, IL18RB, mgC120589, mgC120590

For Research Use Only. Not for use in diagnostic procedures.