missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ IL18R1 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 326) of mouse IL18R1 (NP_032391.1) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 50094M08H250

UNSPSC : 12352202

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13486216

  • £456.00 / 250µg


Estimated Shipment
10-08-2017
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Interleukin 18 receptor 1 (IL18R1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
16182
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
IL18R1, Il18ralpha, Il1rrp
IL18R1
Cytokines & Receptors
His Tag
Lyophilized
36.8kDa
5 x 50μg
Truncated/Recombinant
<1 EU/μg
Interleukin 18 receptor 1
Assay Standard
Mammalian